Learn More
Abnova™ Human C20orf31 Partial ORF (NP_060687, 479 a.a. - 578 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00055741-Q01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
In the endoplasmic reticulum (ER), misfolded proteins are retrotranslocated to the cytosol and degraded by the proteasome in a process known as ER-associated degradation (ERAD). EDEM2 belongs to a family of proteins involved in ERAD of glycoproteins (Mast et al., 2005 [PubMed 15537790]).[supplied by OMIM]
Sequence: PIDPAALHCCQRLKEEQWEVEDLMREFYSLKRCRSKFQKNTVSSGPWEPPARPGTLFSPENHDQARERKPAKQKVPLLSCPSQPFTSKLALLGQVFLDSSEspecificaciones
NP_060687 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
PIDPAALHCCQRLKEEQWEVEDLMREFYSLKRCRSKFQKNTVSSGPWEPPARPGTLFSPENHDQARERKPAKQKVPLLSCPSQPFTSKLALLGQVFLDSS | |
RUO | |
EDEM2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
55741 | |
C20orf31 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
C20orf31/C20orf49/FLJ10783/bA4204.1 | |
EDEM2 | |
Recombinant | |
wheat germ expression system |