Learn More
Abnova™ Human C1orf48 Partial ORF (NP_056286, 182 a.a. - 279 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00025936-Q01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
This gene encodes a protein with two coiled-coil domains that localizes to kinetochores, which are chromosome-associated structures that attach to microtubules and mediate chromosome movements during cell division. The encoded protein is part of a conserved protein complex that includes two chromodomain-containing proteins and a component of the outer plate of the kinetochore. This protein complex is proposed to bridge centromeric heterochromatin with the outer kinetochore structure. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]
Sequence: KEISEAMKSLPALIEQGEGFSQVLRMQPVIHLQRIHQEVFSSCHRKPDAKPENFITQIETTPTETASRKTSDVVLKRKQTKDCPQRKWYPLRPKKINLEspecificaciones
NP_056286 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.52kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
KEISEAMKSLPALIEQGEGFSQVLRMQPVIHLQRIHQEVFSSCHRKPDAKPENFITQIETTPTETASRKTSDVVLKRKQTKDCPQRKWYPLRPKKINL | |
RUO | |
NSL1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
25936 | |
C1orf48 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
C1orf48/DC8/DKFZp566O1646/MIS14 | |
NSL1 | |
Recombinant | |
wheat germ expression system |