missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Bradykinin RB2/BDKRB2 Antibody, R&D Systems™
Mouse Monoclonal Antibody
Marca: R&D Systems MAB47251-100
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Bradykinin RB2/BDKRB2 Monoclonal specifically detects Bradykinin RB2/BDKRB2 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, CyTOF-ready.
Especificaciones
Bradykinin RB2/BDKRB2 | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
B2 bradykinin receptor, B2R, BDKRB2, BK2, BK-2, BK-2 receptor, BKR2, bradykinin receptor B2, BradykininRB2, BRB2, DKFZp686O088 | |
NS0 mouse myeloma cell line transfected with human Bradykinin RB2/BDKRB2, Met1-Gln391, Accession No. P30411 | |
100 μg | |
624.0 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. | |
IgG2b |
Flow Cytometry, Immunohistochemistry, CyTOF | |
471916 | |
Flow Cytometry 0.25 μg/106 cells, Immunohistochemistry 5-25 μg/mL, CyTOF-ready | |
P30411 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
Primary | |
Detects human Bradykinin RB2/BDKRB2 in direct ELISAs. | |
Human | |
Supernatant |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
Human Bradykinin RB2/BDKRB2 Antibody, R&D Systems™ > 100 μg; Unconjgated
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido