missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human AURKC Full-length ORF (NP_001015878.1, 1 a.a. - 309 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00006795-P01.25ug

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16171102

  • 508.00€ / 25 microgramos
Fecha estimada de envío: 12-06-2024
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

This gene encodes a member of the Aurora subfamily of serine/threonine protein kinases. The encoded protein is a chromosomal passenger protein that forms complexes with Aurora-B and inner centromere proteins and may play a role in organizing microtubules in relation to centrosome/spindle function during mitosis. This gene is overexpressed in several cancer cell lines, suggesting an involvement in oncogenic signal transduction. Alternative splicing results in multiple transcript variants. [provided by RefSeq]

Sequence: MSSPRAVVQLGKAQPAGEELATANQTAQQPSSPAMRRLTVDDFEIGRPLGKGKFGNVYLARLKESHFIVALKVLFKSQIEKEGLEHQLRREIEIQAHLQHPNILRLYNYFHDARRVYLILEYAPRGELYKELQKSEKLDEQRTATIIEELADALTYCHDKKVIHRDIKPENLLLGFRGEVKIADFGWSVHTPSLRRKTMCGTLDYLPPEMIEGRTYDEKVDLWCIGVLCYELLVGYPPFESASHSETYRRILKVDVRFPLSMPLGARDLISRLLRYQPLERLPLAQILKHPWVQAHSRRVLPPCAQMAS
Especificaciones

Especificaciones

NP_001015878.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
62kDa
Glutathione Sepharose 4 Fast Flow
25 ug
RUO
AURKC
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, Protein Array, ELISA, Western Blot
6795
AURKC (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
AIE2/AIK3/AurC/STK13/aurora-C
AURKC
Recombinant
wheat germ expression system
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales