Learn More
Abnova™ Human ATG4C Full-length ORF (NP_116241.2, 1 a.a. - 458 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00084938-P01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
Autophagy is the process by which endogenous proteins and damaged organelles are destroyed intracellularly. Autophagy is postulated to be essential for cell homeostasis and cell remodeling during differentiation, metamorphosis, non-apoptotic cell death, and aging. Reduced levels of autophagy have been described in some malignant tumors, and a role for autophagy in controlling the unregulated cell growth linked to cancer has been proposed. This gene encodes a member of the autophagin protein family. The encoded protein is also designated as a member of the C-54 family of cysteine proteases. Alternate transcriptional splice variants, encoding the same protein, have been characterized. [provided by RefSeq]
Sequence: MEATGTDEVDKLKTKFISAWNNMKYSWVLKTKTYFSRNSPVLLLGKCYHFKYEDEDKTLPAESGCTIEDHVIAGNVEEFRKDFISRIWLTYREEFPQIEGSALTTDCGWGCTLRTGQMLLAQGLILHFLGRAWTWPDALNIENSDSESWTSHTVKKFTASFEASLSGEREFKTPTISLKETIGKYSDDHEMRNEVYHRKIISWFGDSPLALFGLHQLIEYGKKSGKKAGDWYGPAVVAHILRKAVEEARHPDLQGITIYVAQDCTVYNSDVIDKQSASMTSDNADDKAVIILVPVRLGGERTNTDYLEFVKGILSLEYCVGIIGGKPKQSYYFAGFQDDSLIYMDPHYCQSFVDVSIKDFPLETFHCPSPKKMSFRKMDPSCTIGFYCRNVQDFKRASEEITKMLKFSSKEKYPLFTFVNGHSRDYDFTSTTTNEEDLFSEDEKKQLKRFSTEEFVLLEspecificaciones
NP_116241.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
78.9kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
RUO | |
ATG4C | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
84938 | |
ATG4C (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
APG4-C/APG4C/AUTL1/AUTL3/FLJ14867 | |
ATG4C | |
Recombinant | |
wheat germ expression system |