Learn More
Abnova™ Human ATG4C Full-length ORF (NP_116241.2, 1 a.a. - 458 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Especificaciones
Número de acceso | NP_116241.2 |
---|---|
Para utilizar con (aplicación) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulación | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
ID de gen (Entrez) | 84938 |
Peso molecular | 78.9kDa |
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
---|---|---|---|---|---|---|---|---|---|
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
16166193
|
Abnova™
H00084938-P01.25UG |
25 ug |
508.00€
25 microgramos |
Fecha estimada de envío: 31-05-2024 Inicie sesión para ver el stock |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | ||||
16156193
|
Abnova™
H00084938-P01.10UG |
10 ug |
335.00€
10 microgramos |
Fecha estimada de envío: 31-05-2024 Inicie sesión para ver el stock |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | ||||
Descripción
Autophagy is the process by which endogenous proteins and damaged organelles are destroyed intracellularly. Autophagy is postulated to be essential for cell homeostasis and cell remodeling during differentiation, metamorphosis, non-apoptotic cell death, and aging. Reduced levels of autophagy have been described in some malignant tumors, and a role for autophagy in controlling the unregulated cell growth linked to cancer has been proposed. This gene encodes a member of the autophagin protein family. The encoded protein is also designated as a member of the C-54 family of cysteine proteases. Alternate transcriptional splice variants, encoding the same protein, have been characterized. [provided by RefSeq]
Sequence: MEATGTDEVDKLKTKFISAWNNMKYSWVLKTKTYFSRNSPVLLLGKCYHFKYEDEDKTLPAESGCTIEDHVIAGNVEEFRKDFISRIWLTYREEFPQIEGSALTTDCGWGCTLRTGQMLLAQGLILHFLGRAWTWPDALNIENSDSESWTSHTVKKFTASFEASLSGEREFKTPTISLKETIGKYSDDHEMRNEVYHRKIISWFGDSPLALFGLHQLIEYGKKSGKKAGDWYGPAVVAHILRKAVEEARHPDLQGITIYVAQDCTVYNSDVIDKQSASMTSDNADDKAVIILVPVRLGGERTNTDYLEFVKGILSLEYCVGIIGGKPKQSYYFAGFQDDSLIYMDPHYCQSFVDVSIKDFPLETFHCPSPKKMSFRKMDPSCTIGFYCRNVQDFKRASEEITKMLKFSSKEKYPLFTFVNGHSRDYDFTSTTTNEEDLFSEDEKKQLKRFSTEEFVLLEspecificaciones
NP_116241.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
78.9kDa | |
Glutathione Sepharose 4 Fast Flow | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
APG4-C/APG4C/AUTL1/AUTL3/FLJ14867 | |
ATG4C | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
84938 | |
ATG4C (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
RUO | |
ATG4C | |
Wheat Germ (in vitro) | |
GST | |
Liquid |