Learn More
Abnova™ Human ASH1L Partial ORF (NP_060959.1, 2859 a.a. - 2967 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00055870-Q01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
This gene encodes a member of the trithorax group of transcriptional activators. The protein contains four AT hooks, a SET domain, a PHD-finger motif, and a bromodomain. It is localized to many small speckles in the nucleus, and also to cell-cell tight junctions. [provided by RefSeq]
Sequence: DALPLIEEVLASQEQAANEIPSLEEPEREGATANVSEGEKKTEESSQEPQSTCTPEERRHNQRERLNQILLNLLEKIPGKNAIDVTYLLEEGSGRKLRRRTLFIPENSFEspecificaciones
NP_060959.1 | |
Liquid | |
55870 | |
ASH1L (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ASH1/ASH1L1/FLJ10504/KIAA1420/KMT2H | |
ASH1L | |
Yes | |
wheat germ expression system |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.73kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
DALPLIEEVLASQEQAANEIPSLEEPEREGATANVSEGEKKTEESSQEPQSTCTPEERRHNQRERLNQILLNLLEKIPGKNAIDVTYLLEEGSGRKLRRRTLFIPENSF | |
RUO | |
ASH1L | |
Wheat Germ (in vitro) | |
GST |