Learn More
Abnova™ Human ASB3 Full-length ORF (AAH06488.1, 1 a.a. - 518 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00051130-P01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
The protein encoded by this gene is a member of the ankyrin repeat and SOCS box-containing (ASB) family of proteins. They contain ankyrin repeat sequence and SOCS box domain. The SOCS box serves to couple suppressor of cytokine signalling (SOCS) proteins and their binding partners with the elongin B and C complex, possibly targeting them for degradation. Multiple alternatively spliced transcript variants have been described for this gene but some of the full length sequences are not known. [provided by RefSeq]
Sequence: MDFTEAYADTCSTVGLAAREGNVKVLRKLLKKGRSVDVADNRGWMPIHEAAYHNSVECLQMLINADSSENYIKMKTFEGFCALHLAASQGHWKIVQILLEAGADPNATTLEETTPLFLAVENGQIDVLRLLLQHGANVNGSHSMCGWNSLHQASFQENAEIIKLLLRKGANKECQDDFGITPLFVAAQYGKLESLSILISSGANVNCQALDKATPLFIAAQEGHTKCVELLLSSGADPDLYCNEDSWQLPIHAAAQMGHTKILDLLIPLTNRACDTGLNKVSPVYSAVFGGHEDCLEILLRNGYSPDAQACLVFGFSSPVCMAFQKDCEFFGIVNILLKYGAQINELHLAYCLKYEKFSIFRYFLRKGCSLGPWNHIYEFVNHAIKAQAKYKEWLPHLLVAGFDPLILLCNSWIDSVSIDTPIFTLEFTNWKTLAPAVERMLSARASNAWILQQHIATVPSLTHLCRLEIRSSLKSERLRSDSYISQLPLPRSLHNYLLYEDVLRMYEVPELAAIQDGEspecificaciones
AAH06488.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
82.72kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
RUO | |
ASB3 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
51130 | |
ASB3 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ASB-3/FLJ10123/FLJ10421/MGC12531/MGC132002/MGC996 | |
ASB3 | |
Recombinant | |
wheat germ expression system |