Learn More
Abnova™ Human ARMCX1 Partial ORF (NP_057692, 188 a.a. - 295 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00051309-Q01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
This gene encodes a member of the ALEX family of proteins and may play a role in tumor suppression. The encoded protein contains a potential N-terminal transmembrane domain and two Armadillo (arm) repeats. Other proteins containing the arm repeat are involved in development, maintenance of tissue integrity, and tumorigenesis. This gene is closely localized with other family members, including ALEX2 and ALEX3, on the X chromosome. [provided by RefSeq]
Sequence: RRGKFNFPYKIDDILSAPDLQKVLNILERTNDPFIQEVALVTLGNNAAYSFNQNAIRELGGVPIIAKLIKTKDPIIREKTYNALNNLSVNAENQGKIKTYISQVCDDTEspecificaciones
NP_057692 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.62kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
RRGKFNFPYKIDDILSAPDLQKVLNILERTNDPFIQEVALVTLGNNAAYSFNQNAIRELGGVPIIAKLIKTKDPIIREKTYNALNNLSVNAENQGKIKTYISQVCDDT | |
RUO | |
ARMCX1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
51309 | |
ARMCX1 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ALEX1/DKFZp686P06199 | |
ARMCX1 | |
Recombinant | |
wheat germ expression system |