Learn More
Abnova™ Human ARL4A Full-length ORF (AAH01111, 1 a.a. - 200 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00010124-P01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
ADP-ribosylation factor-like 4A is a member of the ADP-ribosylation factor family of GTP-binding proteins. ARL4A is similar to ARL4C and ARL4D and each has a nuclear localization signal and an unusually high guaninine nucleotide exchange rate. ARL4A is located in both the nuclear and extranuclear cell compartments. Multiple transcript variants encoding the same protein have been found for this gene. [provided by RefSeq]
Sequence: MGNGLSDQTSILSNLPSFQSFHIVILGLDCAGKTTVLYRLQFNEFVNTVPTKGFNTEKIKVTLGNSKTVTFHFWDVGGQEKLRPLWKSYTRCTDGIVFVVDSVDVERMEEAKTELHKITRISENQGVPVLIVANKQDLRNSLSLSEIEKLLAMGELSSSTPWHLQPTCAIIGDGLKEGLEKLHDMIIKRRKMLRQQKKKREspecificaciones
AAH01111 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
47.74kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
RUO | |
ARL4A | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
10124 | |
ARL4A (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ARL4 | |
ARL4A | |
Recombinant | |
wheat germ expression system |