Learn More
Abnova™ Human AMOT Full-length ORF (NP_573572.1, 1 a.a. - 675 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00154796-P01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
This gene belongs to the motin family of angiostatin binding proteins characterized by conserved coiled-coil domains and C-terminal PDZ binding motifs. The encoded protein is expressed predominantly in endothelial cells of capillaries as well as larger vessels of the placenta where it may mediate the inhibitory effect of angiostatin on tube formation and the migration of endothelial cells toward growth factors during the formation of new blood vessels. Alternative splicing results in multiple transcript variants encoding different isoforms. [provided by RefSeq]
Sequence: MPRAQPSSASYQPVPADPFAIVSRAQQMVEILSDENRNLRQELEGCYEKVARLQKVETEIQRVSEAYENLVKSSSKREALEKAMRNKLEGEIRRMHDFNRDLRERLETANKQLAEKEYEGSEDTRKTISQLFAKNKESQREKEKLEAELATARSTNEDQRRHIEIRDQALSNAQAKVVKLEEELKKKQVYVDKVEKMQQALVQLQAACEKREQLEHRLRTRLERELESLRIQQRQGNCQPTNVSEYNAAALMELLREKEERILALEADMTKWEQKYLEENVMRHFALDAAATVAAQRDTTVISHSPNTSYDTALEARIQKEEEEILMANKRCLDMEGRIKTLHAQIIEKDAMIKVLQQRSRKEPSKTEQLSCMRPAKSLMSISNAGSGLLSHSSTLTGSPIMEEKRDDKSWKGSLGILLGGDYRAEYVPSTPSPVPPSTPLLSAHSKTGSRDCSTQTERGTESNKTAAVAPISVPAPVAAAATAAAITATAATITTTMVAAAPVAVAAAAAPAAAAAPSPATAAATAAAVSPAAAGQIPAAASVASAAAVAPSAAAAAAVQVAPAAPAPVPAPALVPVPAPAAAQASAPAQTQAPTSAPAVAPTPAPTPTPAVAQAEVPASPATGPGPHRLSIPSLTCNPDKTDGPVFHSNTLERKTPIQILGQEPDAEMVEYLIEspecificaciones
NP_573572.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
98.9kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
RUO | |
AMOT | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
154796 | |
AMOT (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
KIAA1071 | |
AMOT | |
Recombinant | |
wheat germ expression system |