missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human AKR7A2 Full-length ORF (AAH10852.1, 1 a.a. - 330 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00008574-P02.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
Aldo-keto reductases, such as AKR7A2, are involved in the detoxification of aldehydes and ketones.[supplied by OMIM]
Sequence: MSRPPPPRVASVLGTMEMGRRMDAPASAAAVRAFLERGHTELDTAFMYSDGQSETILGGLGLGLGGGDCRVKIATKANPWDGKSLKPDSVRSQLETSLKRLQCPQVDLFYLHAPDHGTPVEETLHACQRLHQEGKFVELGLSNYASWEVAEICTLCKSNGWILPTVYQGMYNATTRQVETELFPCLRHFGLRFYAYNPLAGGLLTGKYKYEDKDGKQPVGRFFGNSWAETYRNRFWKEHHFEAIALVEKALQAAYGASAPSVTSAALRWMYHHSQLQGAHGDAVILGMSSLEQLEQNLAATEEGPLEPAVVDAFNQAWHLVAHECPNYFREspecificaciones
AAH10852.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
61.82kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
RUO | |
AKR7A2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
8574 | |
AKR7A2 (Human) Recombinant Protein (P02) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
AFAR/AFAR1/AFB1-AR1/AKR7 | |
AKR7A2 | |
Recombinant | |
wheat germ expression system |