Learn More
Abnova™ Human ABCC6 Full-length ORF (NP_001072996.1, 1 a.a. - 99 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00000368-P01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
The protein encoded by this gene is a member of the superfamily of ATP-binding cassette (ABC) transporters. ABC proteins transport various molecules across extra- and intra-cellular membranes. ABC genes are divided into seven distinct subfamilies (ABC1, MDR/TAP, MRP, ALD, OABP, GCN20, White). The encoded protein, a member of the MRP subfamily, is involved in multi-drug resistance. Mutations in this gene cause pseudoxanthoma elasticum. Alternatively spliced transcript variants that encode different proteins have been described for this gene. [provided by RefSeq]
Sequence: MAAPAEPCAGQGVWNQTEPEPAATSLLSLCFLRTAGVWVPPMYLWVLGPIYLLFIHHHGRGYLRMSPLFKAKMVAAIPGSLEPGNVRGRQGTGWNLVKSEspecificaciones
NP_001072996.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.2kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
RUO | |
ABCC6 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
368 | |
ABCC6 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ABC34/ARA/EST349056/MLP1/MOATE/MRP6/PXE/PXE1 | |
ABCC6 | |
Recombinant | |
wheat germ expression system |