missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HSP60 Antibody (HSPD1/780), Alexa Fluor™ 700, Novus Biologicals™
Mouse Monoclonal Antibody
Marca: Novus Biologicals NBP2-47843AF700
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
HSP60 Monoclonal antibody specifically detects HSP60 in Human, Mouse, Rat, Porcine, Bovine, Canine, Chicken, Hamster, Monkey, Rabbit, Sheep samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Protein Array, Immunofluorescence, CyTOF
Especificaciones
HSP60 | |
Monoclonal | |
Alexa Fluor 700 | |
60 kDa chaperonin, Chaperonin 60, CPN60, GROEL, heat shock 60kD protein 1 (chaperonin), heat shock 60kDa protein 1 (chaperonin), Heat shock protein 60, heat shock protein 65, HLD4, Hsp60, HSP-60, HSP60SPG13, HSP65, HuCHA60, Mitochondrial matrix protein P1,60 kDa heat shock protein, mitochondrial, P60 lymphocyte protein, short heat shock protein 60 Hsp60s1, spastic paraplegia 13 (autosomal dominant) | |
Recombinant human HSP60 protein (Uniprot: P10809) | |
0.1 mL | |
Apoptosis, Cellular Markers, Loading Controls, Membrane Trafficking and Chaperones, Mitochondrial Markers | |
3329 | |
Store at 4°C in the dark. | |
IgG1 κ |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
HSPD1/780 | |
50 mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human, Mouse, Rat, Pig, Bovine | |
Purified |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
HSP60 Antibody (HSPD1/780), Alexa Fluor™ 700, Novus Biologicals™ > 0.1 mL, Alexa Fluor 700
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido