missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HPCL Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Marca: Bio-Techne NBP1-31571
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
HPCL Polyclonal specifically detects HPCL in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Especificaciones
HPCL | |
Polyclonal | |
Unconjugated | |
0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal | |
HACL1 | |
Recombinant protein encompassing a sequence within the C-terminus region of human HACL1. The exact sequence is proprietary. | |
100 μL | |
Primary | |
Tested in human. Not yet tested in any other species. | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
1.0mg/mL | |
Western Blot 1:500-1:3000, Immunohistochemistry 10 - 1:500, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000 | |
1600020H07Rik, 2,2-HPCL, 2-hydroxyacyl-CoA lyase 1, 2-hydroxyphytanoyl-CoA lyase2-hydroxyphytanol-CoA lyase, EC 4.1, HPCL, PHYH2FLJ55041, Phytanoyl-CoA 2-hydroxylase 2, phytanoyl-CoA hydroxylase 2 | |
Rabbit | |
Affinity Purified | |
RUO | |
26061 | |
Human, Mouse | |
IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
HPCL Antibody, Novus Biologicals™ > 0.1mg; Unlabeled
For Research Use Only
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido