missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HLA DRB1 Antibody (HLA-DRB/1067), Alexa Fluor™ 647, Novus Biologicals™
Mouse Monoclonal Antibody
Marca: Novus Biologicals NBP2-47671AF647
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
HLA DRB1 Monoclonal antibody specifically detects HLA DRB1 in Human, Monkey samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Immunofluorescence, CyTOF
Especificaciones
HLA DRB1 | |
Monoclonal | |
Alexa Fluor 647 | |
Clone P2-beta-3, DR1, DR-1, DR12, DR-12, DR13, DR-13, DR14, DR-14, DR16, DR-16, DR4, DR-4, DR5, DR-5, DR7, DR-7, DR8, DR-8, DR9, DR-9, DRB1, DRw10, DRw11, DRw8, DW2.2/DR2.2, FLJ75017, FLJ76359, HLA class II antigen beta chain, HLA class II histocompatibility antigen, DR-1 beta chain, HLA-DR1B, HLA-DRB, HLA-DRB1*, HLA-DRB2, HLA-DR-beta 1, human leucocyte antigen DRB1, leucocyte antigen DR beta 1 chain, leucocyte antigen DRB1, lymphocyte antigen DRB1, major histocompatibility complex, class II, DR beta 1, MHC class II antigen DRB1*1, MHC class II antigen DRB1*10, MHC class II antigen DRB1*11, MHC class II antigen DRB1*12, MHC class II antigen DRB1*13, MHC class II antigen DRB1*14, MHC class II antigen DRB1*15, MHC class II antigen DRB1*16, MHC class II antigen DRB1*3, MHC class II antigen DRB1*4, MHC class II antigen DRB1*7, MHC class II antigen DRB1*8, MHC class II antigen DRB1*9, MHC class II antigen HLA-DR13, MHC class II HLA-DR beta 1 chain, MHC class II HLA-DR-beta cell surface gly | |
Activated human peripheral blood mononuclear cells | |
0.1 mL | |
Asthma, Immunology | |
3123 | |
Store at 4°C in the dark. | |
IgG2b κ |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
HLA-DRB/1067 | |
50 mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human, Monkey | |
Purified |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
HLA DRB1 Antibody (HLA-DRB/1067), Alexa Fluor™ 647, Novus Biologicals™ > 0.1 mL, Alexa Fluor 647
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido