missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HLA-DQ1/DQ3 Monoclonal Antibody (HL-37), Invitrogen™
Mouse Monoclonal Antibody
Marca: Invitrogen MA119143
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Western Blot: non-reducing conditions.
Reacts with polymorphic determinant on human HLA-DQ1 and HLA-DQ3 molecules (recognized epitope was found on isolated b chain of DQ1), but does not react with HLA-DQ2. HLA and MHC antibodies play a significant role in Immunopeptidomics, facilitating the identification and characterization of neoantigens through high-performance liquid chromatography coupled to tandem Mass Spectrometry.
Especificaciones
HLA-DQ1/DQ3 | |
Monoclonal | |
1 mg/mL | |
PBS with 15mM sodium azide; pH 7.4 | |
P01920 | |
HLA-DQB1 | |
Burkitt's lymphoma cell line Raji. | |
100 μg | |
Primary | |
Human, Pig | |
Antibody | |
IgG3 |
Flow Cytometry, Immunoprecipitation, Western Blot | |
HL-37 | |
Unconjugated | |
HLA-DQB1 | |
CELIAC1; HLA class II antigen; HLA class II histocompatibility antigen, DQ beta 1 chain; HLA-D; HLADQ; HLA-DQ; HLADQ1; HLADQ3; HLA-DQB; HLA-DQB1; HLA-DRB1; IDDM1; lymphocyte antigen; major histocompatibility complex, class II, DQ beta 1; MHC Class 2; MHC class II; MHC class II antigen DQB1; MHC class II antigen HLA-DQ-beta-1; MHC class II DQ beta chain; MHC class II HLA-DQ beta glycoprotein; MHC class2 antigen; MHC DQ beta; MHC HLA-D; MHC II HLA | |
Mouse | |
Protein A | |
RUO | |
3119 | |
4° C, do not freeze | |
Liquid |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
HLA-DQ1/DQ3 Monoclonal Antibody (HL-37), Invitrogen™ > 100 μg; Unconjugated
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido