missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HLA-DP Mouse anti-Human, BV421, Clone: B7/21 (also known as B7/21.49; B7/21 (IgG1)), BD Biosciences™
Mouse Monoclonal Antibody
Marca: BD Biosciences 750875
547.77 EUR hasta el 2025-12-31
Use el código promocional "25339" para aplicar la oferta.
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
HLA-DP Monoclonal antibody specifically detects HLA-DP in Human samples. It is validated for Flow Cytometry
Especificaciones
HLA-DP | |
Monoclonal | |
Brilliant Violet 421 | |
HLADP; HLA-DPαβ; DPA and DPB; DPα and DPβ; DPαβ | |
Human NALM6 Leukemia Cells | |
RUO | |
Human | |
IgG1 κ |
Flow Cytometry | |
B7/21 (also known as B7/21.49; B7/21 (IgG1)) | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse | |
50 μg | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions that minimize unconjugated dye and antibody. |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
HLA-DP Mouse anti-Human, BV421, Clone: B7/21 (also known as B7/21.49; B7/21 (IgG1)), BD Biosciences™ > 50 μg; BV421
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido