Learn More
HIV1 Reverse Transcriptase Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA14948
Descripción
PA1-4948 detects the HIV-1 Reverse Transcriptase protein in HIV samples. PA1-4948 has been successfully used in Western blot and ELISA procedures. By Western blot this antibody detects a ∽66 kDa protein and a ∽51 kDa protein, corresponding respectively to the two subunits of HIV-1 reverse transcriptase, in HIV samples. PA1-4948 immunogen corresponds to the HIV-1 Reverse Transcriptase which was over-expressed in E. coli as a recombinant protein. Recombinant HIV-1 Reverse Transcriptase protein (Cat. # RP-4912 or RP-4913) can be purchased for control experiments in Western blot assays.
HIV-1 reverse transcriptase is an RNA-dependent DNA polymerase of HIV-1(AIDS virus), subtype B origin. It also has RNaseH activity and is an enzyme indispensable to the reproduction of AIDS virus.
Especificaciones
HIV1 Reverse Transcriptase | |
Polyclonal | |
Unconjugated | |
HIV RT | |
Full size functional recombinant HIV 1 expressed and purified from E. coli. | |
RUO | |
Virus | |
Antibody | |
IgG |
ELISA, Western Blot | |
Conc. Not Determined | |
whole serum with 0.09% sodium azide | |
Rabbit | |
100 μL | |
Primary | |
-20°C | |
Liquid |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.