Learn More
HDAC2 Mouse anti-Human, CoraLite 594, Clone: 1A3E4, Proteintech
Mouse Monoclonal Antibody
Marca:  Proteintech CL594-67165-100UL
Código de producto. 16801096
  • 450.00€ / 100 microlitros
Descripción
The HDAC2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human HDAC2. This antibody recognizes human, mouse antigen. The HDAC2 antibody has been validated for the following applications: FC (Intra), IF analysis. This antibody is coralite™594.
Especificaciones
Especificaciones
HDAC2
Monoclonal
CoraLite 594
100 μL
3066
IgG2b
Immunofluorescence, Flow Cytometry (Intracellular Staining)
1A3E4
Mouse
Primary
Human, Mouse
Certificados
Promociones
Promociones
Título del producto
HDAC2 Mouse anti-Human, CoraLite 594, Clone: 1A3E4, Proteintech > 100 μL