Learn More
HA Tag Recombinant Mouse Monoclonal Antibody (2-2.2.14), Alexa Fluor™ Plus 647, Invitrogen™
Mouse Recombinant Monoclonal Antibody
Marca: Invitrogen 740005MP647
339.48 EUR hasta el 2025-06-30
Use el código promocional "24108" para aplicar la oferta.
Descripción
HA tag recombinant mouse monoclonal antibody specifically recognizes HA tag at both C and N-terminus of proteins. Alexa Fluor™ Plus recombinant antibodies are conjugated using new, proprietary dye chemistry so you can generate stunning data. Alexa Fluor™ Plus antibodies represent an advancement in fluorescent conjugate technology. Alexa Fluor™ Plus antibodies provide brighter signal compared to leading Alexa Fluor™ antibodies, providing you with better signal-to-noise for your critical experiments. These antibodies show better specificity and lot-to-lot consistency as these are recombinant antibodies, generated by cloning specific genes for the desired antibodies into an expression vector and expressed in vitro. Centrifuge the protein conjugate solution briefly in a microcentrifuge before use; add only the supernatant to the experiment. This step will help eliminate any protein aggregates that may have formed during storage, thereby reducing nonspecific background staining. This 2-2.2.14 antibody has been tested by WB and FC analysis of HEK-293E cells were transiently transfected with HA Tag-protein constructs. This may be used for WB at 1ug/mL , 2.5ug/mL for IHC and for FC at ≤0.5ug per test. Excitation: 658 nm; Emission: 675 nm; Laser: Red Laser Filtration: 0.2um post-manufacturing filtered.
HA Tag Recombinant Monoclonal antibody specifically detects HA Tag in Tag samples. It is validated for Flow Cytometry, Immunocytochemistry, Western Blot
Especificaciones
HA Tag | |
Recombinant Monoclonal | |
1.0 mg/mL | |
Proprietary buffer with 0.008% Bromonitrodioxane, 0.008% Methylisothiazolone; pH 6.8 | |
Mouse | |
Protein A/G | |
RUO | |
Tag | |
Antibody | |
IgG1 |
Flow Cytometry, Immunocytochemistry, Western Blot | |
2-2.2.14 | |
Alexa Fluor Plus 647 | |
HA; HA Epitope Tag; HA Tag; HA-tag; hemagglutinin | |
HA peptide YPYDVPDYA derivitized to ovalbumin. | |
50 μL | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.