missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPAM Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA584441
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Immunogen sequence: DILIIPVGIS YDRIIEGHYN GEQLGKPKKN ESLWSVARGV IRMLRKNYGC VRVDFAQPFS LKEYLESQSQ KPVSALLSLE QALLPAILPS RPSDAADEG
Glycerol-3-phosphate acyltransferase 1 (GPAT1), one of four known GPAT isoforms, is located on the mitochondrial outer membrane, allowing reciprocal regulation with carnitine palmitoyltransferase-1. It is thought to be critical for the development of hepatic steatosis; steatosis triggered by GPAT1 overexpression leads to hepatic and possibly peripheral insulin resistance. GPAT1 is transcriptionally upregulated by insulin and sterol regulatory element binding protein (SREBP-1) and downregulated by AMP-activated protein kinase. Mice deficient in GPAT1 exhibit decreased triacylglycerol (TAG) in cardiomyocytes even in high-fat diets, suggesting that GPAT1 contributes significantly to TAG accumulation in heart tissue during lipogenic or high fat diets. At least two isoforms of GPAT1 are known to exist.
Especificaciones
GPAM | |
Polyclonal | |
Unconjugated | |
Gpam | |
glycerol 3-phosphate acyltransferase, mitochondrial; glycerol-3-phosphate acyltransferase 1, mitochondrial; glycerol-3-phosphate acyltransferase, mitochondrial; GPAM; GPAT; GPAT1; GPAT-1; KIAA1560; P90; RP11-426E5.2 | |
Rabbit | |
Anitgen affinity chromatography, Antigen affinity chromatography | |
RUO | |
57678 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9HCL2 | |
Gpam | |
Recombinant protein corresponding to Human GPAM | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
GPAM Polyclonal Antibody, Invitrogen™ > 100 μL; Unconjugated
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido