missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ GMEB1 Recombinant Protein Antigen
Highly purified. Generating reliable and reproducible results. Applications: Antibody Competition
Marca: Novus Biologicals™ NBP2-55657PEP
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
A recombinant protein antigen with a N-terminal His6-ABP tag corresponding to human GMEB1. Source: E.coli Amino Acid Sequence: STLGNMTTMVSPVELVAMESGLTSAIQAVESTSEDGQTIIEIDPAPDPEAEDTEGKAVILETELRTEEKVVAEMEEHQHQVHNVEI The GMEB1 Recombinant Protein Antigen is derived from E. coli. The GMEB1 Recombinant Protein Antigen has been validated for the following applications: Antibody Competition.
Especificaciones
10691 | |
GMEB1 Recombinant Protein Antigen | |
PBS and 1M Urea, pH 7.4. | |
DNA-binding protein p96PIF, glucocorticoid modulatory element binding protein 1, glucocorticoid modulatory element-binding protein 1, GMEB-1, P96PIF, Parvovirus initiation factor p96, PIF p96, PIF96 | |
Unlabeled | |
100 μl | |
E.Coli |
>80% by SDS-PAGE and Coomassie blue staining | |
Store at −20C. Avoid freeze-thaw cycles. | |
Blocking/Neutralizing, Control | |
GMEB1 | |
Recombinant Protein Antigen | |
RUO | |
This recombinant antigen is only intended to be used as a blocking agent to confirm antibody specificity with the corresponding antibody, catalog number NBP2-51815. It is purified by IMAC chromatography, and the expected concentration is greater than 0.5 mg/ml |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
Novus Biologicals™ GMEB1 Recombinant Protein Antigen > Quantity: 100μL
Para uso exclusivo en investigación
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido