Learn More
Thermo Scientific™ Fluorometric and Luminometric Filter Sets for Varioskan™ ALF Multimode Microplate Reader
Varioskan ALF Multimode Microplate Reader Filter Sets are high quality, validated, and available for various fluorometric and luminometric applications.
Marca: Thermo Scientific™ VAPAI014
Descripción
With the Varioskan ALF Multimode Microplate Reader, fluorescence and certain luminescence measurements are performed with filter-based optics for optimal specificity and sensitivity. The system comes with three pre-installed excitation and emission filter sets suitable for many common fluorometric applications like Quant-iT DNA quantitation assays and PrestoBlue and AlamarBlue cell viability assays. Additionally, five open filter pair positions are available for customized fluorometric and luminometric applications such as measurement of various Alexa Fluor™ and other common dyes or Promega NanoBRET™ assays. The filters are available as predefined, carefully tested, and validated filter sets, to ensure the optimal performance of selected assays.
Varioskan ALF instrument includes the following factory installed fluorometric filter pairs:
- Coumarin family filter pair
- Excitation 345 nm, emission 450 nm
- Fluorescein family filter pair
- Excitation 485, emission 525 nm
- Resorufin family filter pair
- Excitation 555 nm, emission 615 nm
In addition to factory installed filters, a range of validated filter sets are available for various fluorometric and luminometric applications.
- Filter sets consists of high transmission filters for high excitation and emission efficiency.
- Filter sets are designed to efficiently block unwanted light as well as unwanted emitted light to ensure high signal and low background.
- Varioskan ALF filter installation is effortless
- The instrument filter wheel is easy to access, and the correct parameters are quickly set in simple workflow in Thermo Scientific SkanIt software.
- SkanIt software-controlled filter installation process prevents accidental mistakes ensuring error-free

Especificaciones
Filter Set | |
6 to 384-well microplates | |
Each |
Varioskan ALF Multimode Microplate Reader | |
485/575 nm | |
NanoOrange™ Protein Quantitation Kit assays and equivalent dyes |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Para uso en laboratorios