missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ FLT3 (Human) Recombinant Protein
Human FLT3 partial ORF ( NP_004110, 91 a.a. - 190 a.a.) recombinant protein with GST-tag at N-terminal.
Marca: Abnova™ H00002322-Q01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
- Sequence: LQVLVDAPGNISCLWVFKHSSLNCQPHFDLQNRGVVSMVILKMTETQAGEYLLFIQSEATNYTILFTVSIRNTLLYTLRRPYFRKMENQDALVCISESVP
Especificaciones
NP_004110 | |
fms-related tyrosine kinase 3 | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLT3 | |
GST |
2322 | |
Wheat germ expression system | |
10 μg | |
CD135, FLK2, STK1 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |