Learn More
Hellma Flow-Through Cell, 3-in-1, 177-QS
Highly pure Suprasil™ quartz glass flow-through cuvette for absorbance, transmission, and fluorescence measurements and designed with 2 FEP tubing screw connections.
Marca:  Hellma 176-765-85-40
 Ver más versiones de este producto
Código de producto. 15786962
  • 1283.65€ / cada uno
Solo quedan null
Fecha estimada de envoi: 21-11-2025
Descripción
  • High-purity synthetic fused silica quartz construction
  • UV/VIS range
  • Integrated screw connections for FEP tubing
Especificaciones
Especificaciones
110 μL
6 mm
1/Pk
200 - 2500 mm
12.5 mm
Suprasil Quartz
Two Screw Connections, FEB Tubing
10 x 1 mm
35 mm
Documentos
One moment while we fetch your results.
Certificados
Promociones
Promociones
Título del producto
Hellma Flow-Through Cell, 3-in-1, 177-QS > Path length: 10 x 1 mm, Capacity: 110 μL