missing translation for 'onlineSavingsMsg'
Learn More

ACROBiosystems FITC-Labeled Human CEACAM-5 / CD66e Protein, His Tag

MALS verified

383.89€ - 2639.24€

Especificaciones

Peso molecular 72.2 kDa
Fuente FITC-Labeled Human CEACAM-5, His Tag (CE5-HF254) is expressed from human 293 cells (HEK293). It contains AA Lys 35 - Ala 685 (Accession # P06731-1). It is the FITC labeled form of Human CEACAM-5, His Tag (Cat. No.CE5-H5226).
Inmunógeno Lys 35 - Ala 685
Requisitos de almacenamiento -20°C, avoid light
Alias de gen CEACAM-5,CD66e,CEA,Meconium antigen 100
Ver más especificaciones

Productos 2
Código de producto Marca Cantidad Precio Cantidad y disponibilidad  
Código de producto Marca Cantidad Precio Cantidad y disponibilidad  
17196241
Ver documentos Detalles de la promoción
ACROBiosystems
CE5-HF254-25UG
25 μg
383.89€
25 microgramos
Ver productos alternativos
Ahorre hasta 
»
Fecha estimada de envío: 11-04-2024
para ver el stock
Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.
17186241
Ver documentos Detalles de la promoción
ACROBiosystems
CE5-HF254-200UG
200 μg
2639.24€
200 microgramos
Ver productos alternativos
Ahorre hasta 
»
Fecha estimada de envío: 11-04-2024
para ver el stock
Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.
Descripción

Descripción

Carcinoembryonic antigen-related cell adhesion molecule 5 (CEACAM5) is also Carcinoembryonic antigen (CEA), Meconium antigen 100, CD antigen CD66e, CEACAM5 belongs to the immunoglobulin superfamily and CEA family. CEACAM5 contains seven Ig-like (immunoglobulin-like) domains. CEACAM5 is homodimer protein which binding of E.coli Dr adhesins leads to dissociation of the homodimer. CEACAM5 is cell surface glycoprotein that plays a role in cell adhesion and in intracellular signaling. CEACAM5 is receptor for E.coli Dr adhesins.

This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 72.2kDa. The protein migrates as 90-130 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Especificaciones

Especificaciones

72.2 kDa
Lys 35 - Ala 685
CEACAM-5,CD66e,CEA,Meconium antigen 100
FITC
His Tag
Less than 1.0 EU / μg by the LAL method.
Powder
>90% as determined by SDS-PAGE
FITC-Labeled Human CEACAM-5, His Tag (CE5-HF254) is expressed from human 293 cells (HEK293). It contains AA Lys 35 - Ala 685 (Accession # P06731-1). It is the FITC labeled form of Human CEACAM-5, His Tag (Cat. No.CE5-H5226).
-20°C, avoid light
CEACAM-5
Human
PBS, pH7.4
HEK293
Fluorescent labeled protein
Videos
SDS
Documentos

Documentos

Product Certifications