Learn More
ACROBiosystems FITC-Labeled Human CEACAM-5 / CD66e Protein, His Tag
MALS verified
Marca: ACROBiosystems CE5-HF254-200ug
Detalles adicionales : Peso : 0.00010kg
Descripción
Carcinoembryonic antigen-related cell adhesion molecule 5 (CEACAM5) is also Carcinoembryonic antigen (CEA), Meconium antigen 100, CD antigen CD66e, CEACAM5 belongs to the immunoglobulin superfamily and CEA family. CEACAM5 contains seven Ig-like (immunoglobulin-like) domains. CEACAM5 is homodimer protein which binding of E.coli Dr adhesins leads to dissociation of the homodimer. CEACAM5 is cell surface glycoprotein that plays a role in cell adhesion and in intracellular signaling. CEACAM5 is receptor for E.coli Dr adhesins.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 72.2kDa. The protein migrates as 90-130 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Especificaciones
72.2 kDa | |
FITC-Labeled Human CEACAM-5, His Tag (CE5-HF254) is expressed from human 293 cells (HEK293). It contains AA Lys 35 - Ala 685 (Accession # P06731-1). It is the FITC labeled form of Human CEACAM-5, His Tag (Cat. No.CE5-H5226). | |
-20°C, avoid light | |
CEACAM-5 | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Fluorescent labeled protein |
200 μg | |
Lys 35 - Ala 685 | |
CEACAM-5,CD66e,CEA,Meconium antigen 100 | |
FITC | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE |