missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACROBiosystems FITC-Labeled Human CD72 Protein, Fc Tag
387.09€ - 2661.24€
Especificaciones
Número de acceso | AAH30227 |
---|---|
Peso molecular | 54.2 kDa |
Fuente | FITC-Labeled Human CD72, Fc Tag (CD2-HF258) is expressed from human 293 cells (HEK293). It contains AA Arg 117 - Asp 359 (Accession # P21854-1). It is the FITC labeled form of Human CD72, Fc Tag (Cat. No.CD2-H5251). |
Inmunógeno | Arg 117 - Asp 359 |
Requisitos de almacenamiento | -20°C, avoid light |
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
---|---|---|---|---|---|---|---|---|---|
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
17196111
|
ACROBiosystems
CD2-HF258-25UG |
25 μg |
387.09€
25 microgramos |
Fecha estimada de envío: 02-05-2024 Inicie sesión para ver el stock |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | ||||
17186111
|
ACROBiosystems
CD2-HF258-200UG |
200 μg |
2661.24€
200 microgramos |
Fecha estimada de envío: 02-05-2024 Inicie sesión para ver el stock |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | ||||
Descripción
B-cell differentiation antigen CD72 is also known as Lyb-2. CD72 contains one C-type lectin domain. CD72 is a protein active in the immune system of animals. CD72 consists of two identical halves, each of about 39-43 kD, and is a C-type lectin. Its primarily locus of expression is B-cells, where it appears to mediate aspects of B-cell - T-cell interaction. CD72 plays a role in B-cell proliferation and differentiation. CD72 is a ligand for CD5.
This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 54.2 kDa. The protein migrates as 60-65 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Especificaciones
AAH30227 | |
FITC-Labeled Human CD72, Fc Tag (CD2-HF258) is expressed from human 293 cells (HEK293). It contains AA Arg 117 - Asp 359 (Accession # P21854-1). It is the FITC labeled form of Human CD72, Fc Tag (Cat. No.CD2-H5251). | |
-20°C, avoid light | |
CD72 | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Fluorescent labeled protein |
54.2 kDa | |
Arg 117 - Asp 359 | |
CD72,Lyb-2,Lyb2 | |
FITC | |
Fc Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE |