Learn More
ACROBiosystems FITC-Labeled Human CD72 Protein, Fc Tag
Marca:  ACROBiosystems CD2-HF258-25ug
Detalles adicionales : Peso : 0.00010kg
 Ver más versiones de este producto
Código de producto. 17196111
  • 363.80€ / 25 microgramos
Fecha estimada de envío: 09-01-2023
para ver el stock
Descripción
Descripción
B-cell differentiation antigen CD72 is also known as Lyb-2. CD72 contains one C-type lectin domain. CD72 is a protein active in the immune system of animals. CD72 consists of two identical halves, each of about 39-43 kD, and is a C-type lectin. Its primarily locus of expression is B-cells, where it appears to mediate aspects of B-cell - T-cell interaction. CD72 plays a role in B-cell proliferation and differentiation. CD72 is a ligand for CD5.
This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 54.2 kDa. The protein migrates as 60-65 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Especificaciones
Especificaciones
AAH30227
54.2 kDa
25 µg
Arg 117 - Asp 359
CD72,Lyb-2,Lyb2
FITC Labeled Protein
PBS, pH7.4
HEK293
Human
Powder
>90% as determined by SDS-PAGE
FITC-Labeled Human CD72, Fc Tag (CD2-HF258) is expressed from human 293 cells (HEK293). It contains AA Arg 117 - Asp 359 (Accession # P21854-1). It is the FITC labeled form of Human CD72, Fc Tag (Cat. No.CD2-H5251).
-20°C, avoid light
CD72
Fc Tag
Less than 1.0 EU / µg by the LAL method.
Fluorescent labeled protein