Learn More
Fibroblast activation protein alpha Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 27596-1-AP-20UL
Descripción
The Fibroblast activation protein alpha antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human Fibroblast activation protein alpha. This antibody recognizes human, mouse antigen. The Fibroblast activation protein alpha antibody has been validated for the following applications: IHC, ELISA analysis.
Especificaciones
Fibroblast activation protein alpha | |
500 μg/mL | |
Rabbit | |
Primary | |
Human, Mouse | |
150 kDa | |
IgG |
Immunohistochemistry, ELISA | |
DPPIV, FAP, FAPA, Seprase | |
20 μL | |
2191 | |
Store at -20°C. Stable for one year after shipment. | |