Learn More
FCGR2A / CD32a Mouse anti-Human, CoraLite 488, Clone: 1D3F1, Proteintech
Mouse Monoclonal Antibody
Marca: Proteintech CL488-66529-100UL
Descripción
The FCGR2A / CD32a antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human FCGR2A / CD32a. This antibody recognizes human antigen. The FCGR2A / CD32a antibody has been validated for the following applications: FC, IF analysis. This antibody is coralite™488.
Especificaciones
FCGR2A / CD32a | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human |
Immunofluorescence, Flow Cytometry | |
1D3F1 | |
CoraLite 488 | |
100 μL | |
2212 | |
IgG2b |