missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ F(ab')2-Donkey anti-Mouse IgG (H+L) Highly Cross-Adsorbed Secondary Antibody, FITC
F(ab')2-Donkey Polyclonal Secondary Antibody
Marca: Invitrogen™ A24507
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
F(ab')2-Donkey anti-Mouse IgG (H+L) Highly Cross-Adsorbed Secondary Antibody for ICC/IF, Flow
Anti-Mouse secondary antibodies are affinity-purified antibodies with well-characterized specificity for mouse immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Especificaciones
Mouse IgG (H+L) Highly Cross-Adsorbed | |
Polyclonal | |
FITC | |
Donkey | |
Affinity Chromatography | |
RUO | |
Mouse | |
Secondary Antibody | |
IgG |
Flow Cytometry, Immunocytochemistry | |
2.5 mg/mL | |
PBS with 1% BSA and 0.05% sodium azide; pH 7.8 | |
Gamma Immunoglobins Heavy and Light chains. | |
500 μg | |
Secondary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark. | |
Liquid |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
Invitrogen™ F(ab')2-Donkey anti-Mouse IgG (H+L) Highly Cross-Adsorbed Secondary Antibody, FITC > 500 μg; FITC
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido