Learn More
Invitrogen™ Thiobarbituric Acid Reactants (TBARS) Colorimetric Assay Kit
These complete, ready-to-use kits include clear 96-well plate(s), other assay specific reagents, buffers, and a lot-specific technical data sheet.
Marca: Invitrogen™ EEA021
Descripción
The Thiobarbituric Acid Reactants (TBARS) Colorimetric Assay Kit is a highly reliable and sensitive assay designed for the quantitative measurement of TBARS in serum, plasma, animal tissue, cultured cells, and other sample types. TBARS are products of lipid peroxidation, a process that occurs when polyunsaturated fatty acids in cellular membranes are attacked by oxygen free radicals. TBARS, particularly malondialdehyde (MDA), are commonly used as biomarkers of lipid peroxidation and can reflect the degree of oxidative stress and cellular injury in organisms.
In this assay, TBARS and thiobarbituric acid (TBA) react under high temperature and acidic conditions. This reaction forms a pink compound, which has an optical density measured at 530–540 nm and the concentration of which is linearly related to the concentration of TBARS in the sample.
Key features
• Simple and fast protocol—user-friendly, requiring minimal hands-on time and providing rapid results within a short incubation period
• Broad range of kits—across key analytes and targets within the most important metabolic pathways
• Consistent—strict quality control criteria to ensure reproducible and reliable results

Especificaciones
2.6–100 μmol/L | |
Store at 2–8°C. |
|
Plasma, Serum, Tissue Homogenates | |
60 min. | |
4.3 | |
100 μl | |
Thiobarbituric Acid Reactants (TBARS) |
0.85 μmol/L | |
Colorimétrico | |
Lector de microplacas | |
7.1 | |
96 pruebas | |
Dry Ice, 2–8°C | |
80 min. |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.