missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dengue Virus Type 2 prM Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Thermo Scientific PA5117434
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Centrifuge briefly prior to opening.
Dengue fever and dengue hemorrhagic fever are acute febrile diseases, found in the tropics and Africa. Caused by one of four closely related virus serotypes of the genus Flavivirus, family Flaviviridae, each serotype is sufficiently different that there is no cross-protection and epidemics caused by multiple serotypes (hyperendemicity) can occur. Dengue is transmitted to humans by the Aedes aegypti (rarely Aedes albopictus) mosquito, which feeds during the day.
Especificaciones
Dengue Virus Type 2 prM | |
Polyclonal | |
Unconjugated | |
Dengue 2 prm; Dengue prm; PrM protein | |
Recombinant protein encompassing a sequence within the center region of Dengue virus prM protein. The exact sequence is proprietary. | |
100 μL | |
Primary | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
1.33 mg/mL | |
PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
Virus | |
Antibody | |
IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
Dengue Virus Type 2 prM Polyclonal Antibody, Invitrogen™ > 100 μL; Unconjugated
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido