missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Human CSK, His Tag Recombinant Protein
Human CSK, His Tag Recombinant Protein
Marca: Thermo Scientific™ PV4075
Detalles adicionales : Peso : 0.05250kg
Descripción
- CSK is a tyrosine kinase that has been shown to downregulate the activity of SRC oncoprotein through phosphorylation of its carboxy terminus
- Since cell transformation by SRC is caused by various mechanisms that interfere with this phosphorylation, the CSK gene might function as an antioncogene
Especificaciones
NP_004374 | |
51.9 kDa | |
CSK | |
Human | |
His Tag | |
Tyrosine Kinases | |
Full Length | |
Drug Development |
Kinase Assay | |
1 mg | |
CSK Kinase Family | |
Recombinant | |
Storage Buffer: 50 mM Tris, pH 7.5, 100 mM NaCl, 10% glycerol Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
|
Dry Ice | |
Frozen |