missing translation for 'onlineSavingsMsg'
Learn More
Learn More
COL1A1 Monoclonal Antibody (C1), Invitrogen™
Mouse Monoclonal Antibody
Marca: Invitrogen MA550846
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
The protein encoded by this gene is the pro-alpha1 chain of type I collagen whose triple helix comprises two alpha1 chains and one alpha2 chain. Type I is a fibril-forming collagen found in most connective tissues and is abundant in bone, cornea, dermis and tendon. Mutations in this gene are associated with osteogenesis imperfecta types I-IV, Ehlers-Danlos syndrome type VIIA, Ehlers-Danlos syndrome Classical type, Caffey Disease and idiopathic osteoporosis. Reciprocal translocations between chromosomes 17 and 22, where this gene and the gene for platelet-derived growth factor beta are located, are associated with a particular type of skin tumor called dermatofibrosarcoma protuberans, resulting from unregulated expression of the growth factor. Two transcripts, resulting from the use of alternate polyadenylation signals, have been identified for this gene.
Especificaciones
COL1A1 | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.05% Proclin 300, pH 7.4 | |
P02452 | |
COL1A1 | |
Recombinant Human COL1a1 protein, Arg253-Ala471 (Accession #P02452) | |
20 μL | |
Primary | |
Human, Pig | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
C1 | |
Unconjugated | |
COL1A1 | |
alpha-1 type 1 collagen, alpha-1 type I collagen, alpha1(I) procollagen, COL1A, Col1a1, Col1a-1, Cola1, Cola-1, COLIA1, Collagen 1, collagen alpha 1 chain type I, collagen alpha-1(I) chain, collagen alpha-1(I) chain preproprotein, collagen of skin, tendon and bone, alpha-1 chain, collagen type I alpha 1, collagen, type 1, alpha 1, collagen, type I, alpha 1, Collagen1A1, EDSC, Mov13, Mov-13, OI1, OI2, OI3, OI4, pro-alpha-1 collagen type 1, procollagen type I, alpha 1, procollagen, type 1, alpha 1, procollagen, type I, alpha 1, Type I Collagen, type I proalpha 1, type I procollagen alpha 1 chain, type I-alpha 1 collagen | |
Mouse | |
Protein A/G | |
RUO | |
100738123, 1277 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
IgG1 κ |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
COL1A1 Monoclonal Antibody (C1), Invitrogen™ > 20 μL; Unconjugated
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido