missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Claudin 18 Monoclonal Antibody (2F8), Invitrogen™
Mouse Monoclonal Antibody
Marca: Invitrogen MA550294
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Claudin-18 is a tight junction protein expressed as lung-and stomach-specific isoforms which are generated by alternative splicing of the claudin-18 gene. The lung-specific form is a downstream target gene regulated by the T/EBP/NKX2.1 transcription factor. A splice variant lacking the C-terminal cytoplasmic domain also exists in mouse, but has not been confirmed in human. Human claudin-18 demonstrates 88% amino acid sequence identity to the mouse protein. Immunohistochemical studies have demonstrated complete membrane localization of claudin-18 in lung and stomach epithelial cells, while electron microscopy has shown that it is concentrated in the cell-cell borders of these cells. These features suggest a potentially important role for claudin-18 in the structure and function of tight junctions in the lung and stomach. Claudin-18 expression has also been reported in the inner ear.
Especificaciones
Claudin 18 | |
Monoclonal | |
1 mg/mL | |
PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide | |
P56856 | |
Cldn18 | |
Synthesized peptide derived from human Claudin 18 | |
100 μL | |
Primary | |
Human | |
Liquid |
ELISA, Immunohistochemistry (Paraffin) | |
2F8 | |
Unconjugated | |
Cldn18 | |
claudin 18, claudin 18-like, claudin-18, CLDN18, SFTA5, SFTPJ, surfactant associated 5, surfactant associated protein J, surfactant, pulmonary associated protein J, UNQ778/PRO1572 | |
Mouse | |
Affinity chromatography | |
RUO | |
51208 | |
-20°C or -80°C if preferred | |
IgG1 λ |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
Claudin 18 Monoclonal Antibody (2F8), Invitrogen™ > 100 μL; Unconjugated
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido