missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Claudin 11 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
121.38€ - 325.38€
Especificaciones
Antígeno | Claudin 11 |
---|---|
Concentración | 0.27 mg/mL |
Aplicaciones | Immunohistochemistry (Paraffin), Western Blot, Flow Cytometry, Immunofluorescence, Immunocytochemistry |
Clasificación | Polyclonal |
Conjugado | Unconjugated |
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
---|---|---|---|---|---|---|---|---|---|
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
16819693
|
Proteintech
12152-1-AP-20UL |
20 μL |
Este artículo actualmente no está disponible o ha sido discontinuado. Ver la página del producto para posibles alternativas |
||||||
16809693
|
Proteintech
12152-1-AP-150UL |
150 μL |
Este artículo actualmente no está disponible o ha sido discontinuado. Ver la página del producto para posibles alternativas |
||||||
Descripción
The claudin superfamily consists of many structurally related proteins in humans. These proteins, which include claudin 1 through 18, are important structural and functional components of tight junctions in paracellular transport. Claudins are located in both epithelial and endothelial cells in all tight junction-bearing tissues. Claudins, which consist of four trans-membrane domains and two extracellular loops, make up tight junction strands. Claudin-11 is an oligodendrocyte-specific protein that is expressed in the tight junctions of Sertoli cells and myelin sheaths in mice.Especificaciones
Claudin 11 | |
Immunohistochemistry (Paraffin), Western Blot, Flow Cytometry, Immunofluorescence, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
Human | |
O75508 | |
5010 | |
Claudin 11 Fusion Protein Ag2764 | |
Primary | |
-20°C | |
CLDN11 |
0.27 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
claudin 11, CLDN11, OSP, OTM | |
CLDN11 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |