missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Claudin 11 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 12152-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
The claudin superfamily consists of many structurally related proteins in humans. These proteins, which include claudin 1 through 18, are important structural and functional components of tight junctions in paracellular transport. Claudins are located in both epithelial and endothelial cells in all tight junction-bearing tissues. Claudins, which consist of four trans-membrane domains and two extracellular loops, make up tight junction strands. Claudin-11 is an oligodendrocyte-specific protein that is expressed in the tight junctions of Sertoli cells and myelin sheaths in mice.Especificaciones
Claudin 11 | |
Polyclonal | |
Unconjugated | |
CLDN11 | |
claudin 11, CLDN11, OSP, OTM | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
5010 | |
-20°C | |
Liquid |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O75508 | |
CLDN11 | |
Claudin 11 Fusion Protein Ag2764 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |