missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CHI3L1 Recombinant Rabbit Monoclonal Antibody (101), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Marca: Invitrogen MA543079
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Specificity: Human CHI3L1/YKL40
Human cartilage glycoprotein 39 (GP-39), also known as YKL-40, is a glycoprotein secreted by articular chondrocytes, synoviocytes and macrophages. Serum and synovial fluid GP-39 levels are elevated in inflammatory diseases and correlate with the degree of joint destruction in rheumatoid arthritis. GP-39 is expressed in articular chondrocytes and synovial cells, as well as in liver, but is undetectable in muscle tissues, lung, pancreas, mononuclear cells and fibroblasts. GP-39 is a candidate autoantigen in rheumatoid arthritis and is important in the capacity of cells to respond to and cope with changes in their environment.
Especificaciones
CHI3L1 | |
Recombinant Monoclonal | |
1 mg/mL | |
PBS with no preservative | |
P36222 | |
CHI3L1 | |
Recombinant Human CHI3L1/YKL40 protein. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
ELISA | |
101 | |
Unconjugated | |
CHI3L1 | |
38 kDa heparin-binding glycoprotein; 38kDa heparin-binding glycoprotein; 39 kDa synovial protein; 39 kDa whey protein; 40 kDa secretory signalling glycoprotein; ASRT7; AW208766; BP40; BP40 precursor; breast regression protein 39; Brp39; BRP39 protein; Cartilage glycoprotein 39; cartilage glycoprotein-39; CGP39; CGP-39; CHI3L1; Chil1; chitinase 3 like 1; Chitinase 3 like protein 1CGP 39; chitinase 3-like 1; chitinase 3-like 1 (cartilage glycoprotein-39); chitinase 3-like 1 protein; chitinase-3-like protein 1; chitinase-like 1; chitinase-like protein 1; CLP-1; DKFZp686N19119; FLJ38139; GP38K; Gp39; GP-39; hCGP39; HC-gp39; hCGP-39; HCGP-3P; Mammary gland protein 40; MGP-40; signal processing protein; signal-processing protein; SPC-40; SPS-40; YKL40; YKL-40; YYL-40 | |
Rabbit | |
Protein A | |
RUO | |
1116 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
CHI3L1 Recombinant Rabbit Monoclonal Antibody (101), Invitrogen™ > 100 μL; Unconjugated
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido