missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CENPO Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 20611-1-AP-20UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Accurate chromosome segregation during mitosis requires a kinetochore to assemble correctly at the centromere of each chromatid and form a dynamic interface with the microtubules of the mitotic spindle. The kinetochore assembly includes the multisubunit CENP-H/I complex which can be divided into three functional classes based on phenotype analysis. One of these classes is referred to as the CENP-O class of proteins of which CENPO is the exemplar. These proteins form a stable complex with each other and are required for proper kinetochore function and for recovery from mitotic spindle damage. CENPO has also been shown to be an auto-antigen in a small population of anti-centromere antibodies- (ACA-) positive patients with sclerodoma.Especificaciones
CENPO | |
Polyclonal | |
Unconjugated | |
CENPO | |
CENP O, CENPO, centromere protein O, ICEN36, MCM21R | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
52504, 684439, 79172 | |
-20°C | |
Liquid |
Western Blot | |
0.35 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8K015, Q9BU64 | |
Cenpo | |
CENPO Fusion Protein Ag14619 | |
20 μL | |
Primary | |
Mouse, Human, Rat | |
Antibody | |
IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
CENPO Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 20 μL; Unconjugated
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido