missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ CDKN1B (Human) Recombinant Protein
Human CDKN1B full-length ORF ( AAH01971, 1 a.a. - 198 a.a.) recombinant protein with GST-tag at N-terminal.
Marca: Abnova™ H00001027-P01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
- Sequence: MSNVRVSNGSPSLERMDARQAEHPKPSACRNLFGPVDHEELTRDLEKHCRDMEEASQRKWNFDFQNHKPLEGKYEWQEVEKGSLPEFYYRPPRPPKGACKVPAQESQDGSGSRPAAPLIGAPANSEDTHLVDPKTDPSDSQTGLAEQCAGIRKRPATGDSSTQNKRANRTEENVSDGSPNAGSVEQTPKKPGLRRRQT
Especificaciones
AAH01971 | |
cyclin-dependent kinase inhibitor 1B (p27, Kip1) | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CDKN1B | |
GST |
1027 | |
Wheat germ expression system | |
25 μg | |
CDKN4, KIP1, MEN1B, MEN4, P27KIP1 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |