missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD91 Monoclonal Antibody (A2MR-a2), Super Bright™ 600, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Especificaciones
Antígeno | CD91 |
---|---|
Clon | A2MR-a2 |
Concentración | 5 μL/Test |
Aplicaciones | Flow Cytometry |
Clasificación | Monoclonal |
Descripción
Mouse Monoclonal Antibody
The protein encoded by this gene is an endocytic receptor involved in several cellular processes, including intracellular signaling, lipid homeostasis, and clearance of apoptotic cells. In addition, the encoded protein is necessary for the A2M-mediated clearance of secreted amyloid precursor protein and beta-amyloid, the main component of amyloid plaques found in Alzheimer patients. Expression of this gene decreases with age and has been found to be lower than controls in brain tissue from Alzheimer patients.Especificaciones
CD91 | |
5 μL/Test | |
Monoclonal | |
Liquid | |
RUO | |
PBS with BSA and 0.09% sodium azide; pH 7.2 | |
4035 | |
IgG1 κ | |
Affinity chromatography | |
Antibody |
A2MR-a2 | |
Flow Cytometry | |
Super Bright 600 | |
Mouse | |
Human | |
Q07954 | |
LRP1 | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
LRP1 |
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
CD91 Monoclonal Antibody (A2MR-a2), Super Bright™ 600, eBioscience™, Invitrogen™