missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD90.2 (Thy-1.2) Monoclonal Antibody (53-2.1), NovaFluor™ Blue 610-70S, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
Marca: Invitrogen M028T02B06
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
CD90.2 (Thy 1) antigen is a GPI linked glycoprotein member of the Immunoglobulin super family. CD90.2 is expressed in murine T cells, thymocytes, neural cells, Kupffer's cells and fibroblasts. Thy 1 may play a role in cell-cell or cell-ligand interactions during synaptogenesis and other events in the brain. CD90.2 is an 18 kDa plasma membrane antigen expressed by hematopoietic cells and neurons in all species studied, highly expressed in connective tissue, various fibroblast and stromal cell lines and lastly, by tumor endothelial cell lines. In the mouse, CD90.2 is expressed on all thymocytes and peripheral T lymphocytes.
Especificaciones
CD90.2 (Thy-1.2) | |
Monoclonal | |
0.1 mg/mL | |
PBS with 0.09% sodium azide; pH 7.2 | |
P01831 | |
Thy1 | |
Affinity chromatography | |
RUO | |
21838 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
53-2.1 | |
NovaFluor Blue 610-70S | |
Thy1 | |
CD90; FLJ33325; T25; Thy 1.2; Thy1; Thy-1; Thy-1 antigen; thy-1 membrane glycoprotein; Thy1.1; Thy1.2; Thy-1.2; thymus cell antigen 1, theta | |
Rat | |
25 μg | |
Primary | |
Mouse | |
Antibody | |
IgG2a κ |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
CD90.2 (Thy-1.2) Monoclonal Antibody (53-2.1), NovaFluor™ Blue 610-70S, eBioscience™, Invitrogen™ > 25 μg; NovaFluor Blue 610-70S
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido