Learn More
CD8a Rat, anti-Mouse, FITC, Clone: 53-6.7, STEMCELL Technologies
Rat Monoclonal Antibody
98.42€ - 340.48€
Especificaciones
Antígeno CD8a
Clon 53-6.7
Aplicaciones Functional Assay, Immunofluorescence, Immunocytochemistry, Flow Cytometry, CyTOF
Clasificación Monoclonal
Conjugado FITC
Ver más especificaciones
Productos 2
Código de producto Marca Cantidad Precio Cantidad y disponibilidad  
Código de producto Marca Cantidad Precio Cantidad y disponibilidad  
17188551
Ver documentos Detalles de la promoción
Stemcell Technologies
60023FI
500 μg
340.48€
500 microgramos
Ver productos alternativos
Ahorre hasta 
»
Fecha estimada de envío: 15-12-2022
para ver el stock
17198551
Ver documentos Detalles de la promoción
Stemcell Technologies
60023FI.1
50 μg
98.42€
50 microgramos
Ver productos alternativos
Ahorre hasta 
»
Fecha estimada de envío: 15-12-2022
para ver el stock
Descripción
Descripción
  • The 53-6.7 antibody reacts with murine CD8a, a 32 - 34 kDa type I transmembrane glycoprotein which is a subunit of CD8
  • CD8 is a disulfide-bonded dimer, found either as a heterodimer of CD8a (α) and CD8b (β) subunits (i.e., αβ) or a homodimer (αα)
  • CD8 acts as a co-receptor to the T cell receptor (TCR) during T cell activation by binding MHC Class I molecules presented by an antigen-presenting cell
  • It functions to strengthen the association between the TCR and MHC I-antigen complex and to amplify signals from the TCR to the cytoplasm through the interaction of its intracellular domain with cytoplasmic tyrosine kinases such as Lck
  • The CD8a chain binds to the alpha-3 domain of class I MHC molecules
  • This antibody clone has been verified for purity assessments of cells isolated with EasySep™ kits, including EasySep™ Mouse CD8+ T Cell Isolation Kit (Catalog #19853) and EasySep™ Mouse CD90.2 Positive Selection Kit II (Catalog #18951)
Especificaciones
Especificaciones
CD8a
Functional Assay, Immunofluorescence, Immunocytochemistry, Flow Cytometry, CyTOF
FITC
Frog, Mouse
Primary
53-6.7
Monoclonal
Rat
IgG2a κ