Learn More
CD8a Rat, anti-Mouse, FITC, Clone: 53-6.7, STEMCELL Technologies
Rat Monoclonal Antibody
Marca: Stemcell Technologies 60023FI
Detalles adicionales : Peso : 0.00110kg
Descripción
- The 53-6.7 antibody reacts with murine CD8a, a 32 - 34 kDa type I transmembrane glycoprotein which is a subunit of CD8
- CD8 is a disulfide-bonded dimer, found either as a heterodimer of CD8a (α) and CD8b (β) subunits (i.e., αβ) or a homodimer (αα)
- CD8 acts as a co-receptor to the T cell receptor (TCR) during T cell activation by binding MHC Class I molecules presented by an antigen-presenting cell
- It functions to strengthen the association between the TCR and MHC I-antigen complex and to amplify signals from the TCR to the cytoplasm through the interaction of its intracellular domain with cytoplasmic tyrosine kinases such as Lck
- The CD8a chain binds to the alpha-3 domain of class I MHC molecules
- This antibody clone has been verified for purity assessments of cells isolated with EasySep™ kits, including EasySep™ Mouse CD8+ T Cell Isolation Kit (Catalog #19853) and EasySep™ Mouse CD90.2 Positive Selection Kit II (Catalog #18951)
Especificaciones
CD8a | |
Monoclonal | |
FITC | |
500 μg | |
Mouse, Frog | |
T Cells, T Cells, CD8+ |
CyTOF, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Functional Assay | |
53-6.7 | |
Rat | |
Primary | |
IgG2a κ |