missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD88 Rat anti-Mouse, Oligo, Clone: 20/70, BD™ AbSeq ™
Rat Monoclonal Antibody
Marca: BD Biosciences 940438
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
CD88 Monoclonal antibody specifically detects CD88 in Mouse samples. It is validated for Single Cell 3' Sequencing
Especificaciones
CD88 | |
Monoclonal | |
Oligo | |
Cd88; C5ar1; C5a-R; C5aR; C5r1; C5a Ligand; C5a anaphylatoxin receptor | |
Mouse C5aR Transfected Cell Line | |
RUO | |
Mouse | |
IgG2b κ |
Single Cell 3' Sequencing | |
Clone 20/70 | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Rat | |
25 Tests | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography and conjugated to BD AbSeq oligonucleotide under optimal conditions. |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
CD88 Rat anti-Mouse, Oligo, Clone: 20/70, BD™ AbSeq ™ > 25 Tests; Oligo
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido