missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD85g (ILT7) Mouse anti-Human, BUV737, Clone: 17G10.2 (also known as 17.2), BD Biosciences™
Mouse Monoclonal Antibody
Marca: BD Biosciences 749344
391.68 EUR hasta el 2025-12-31
Use el código promocional "25339" para aplicar la oferta.
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
CD85g (ILT7) Monoclonal antibody specifically detects CD85g (ILT7) in Human samples. It is validated for Flow Cytometry
Especificaciones
CD85g (ILT7) | |
Monoclonal | |
Brilliant Ultraviolet 737 | |
LILRA4; CD85g; ILT7; ILT-7; Immunoglobulin-like transcript 7 | |
ILT7–FcεRIγ-transfected 2B4 cells | |
RUO | |
Human | |
IgG1 κ |
Flow Cytometry | |
17G10.2 (also known as 17.2) | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse | |
50 μg | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon BUV737 under optimal conditions that minimize unconjugated dye and antibody. |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
CD85g (ILT7) Mouse anti-Human, BUV737, Clone: 17G10.2 (also known as 17.2), BD Biosciences™ > 50 μg; BUV737
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido