missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD71 Rat anti-Mouse, BUV737, Clone: C2 (also known as C2F2), BD Biosciences™
Rat Monoclonal Antibody
Marca: BD Biosciences 741718
441.74 EUR hasta el 2025-12-31
Use el código promocional "25339" para aplicar la oferta.
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
CD71 Monoclonal antibody specifically detects CD71 in Mouse samples. It is validated for Flow Cytometry
Especificaciones
CD71 | |
Monoclonal | |
Brilliant Ultraviolet 737 | |
Transferrin Receptor; TR; TfR; TfR1; Tfrc; Trfr; Mtvr-1 | |
Mouse cell line | |
RUO | |
Mouse | |
IgG1 κ |
Flow Cytometry | |
C2 (also known as C2F2) | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Rat | |
50 μg | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon BUV737 under optimal conditions that minimize unconjugated dye and antibody. |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
CD71 Rat anti-Mouse, BUV737, Clone: C2 (also known as C2F2), BD Biosciences™ > 50 μg; BUV737
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido